MilliporeSigma
  • Degradation of 2-bromo-, 2-chloro- and 2-fluorobenzoate by Pseudomonas putida CLB 250.
Degradation of 2-bromo-, 2-chloro- and 2-fluorobenzoate by Pseudomonas putida CLB 250.
FEMS microbiology letters (1989-07-15)
K H Engesser, P Schulte
ABSTRACT
Pseudomonas putida strain CLB 250 (DSM 5232) utilized 2-bromo-, 2-chloro- and 2-fluorobenzoate as sole source of carbon and energy. Degradation is suggested to be initiated by a dioxygenase liberating halide in the first catabolic step. After decarboxylation and rearomatization catechol is produced as a central metabolite which is degraded via the ortho-pathway. After inhibition of ring cleavage activities with 3-chlorocatechol, 2-chlorobenzoate was transformed to catechol in nearly stoichiometric amounts. Other ortho-substituted benzoates like anthranilate and 2-methoxybenzoate seem to be metabolized via the same route.
Sigma-Aldrich
2-Bromobenzoic acid, 97%
Sigma-Aldrich
2-Fluorobenzoic acid, 97%