Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13284

Supelco

3-Methoxypropylamine

analytical standard

Synonym(s):

1-Amino-3-methoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2)3NH2
CAS Number:
Molecular Weight:
89.14
Beilstein/REAXYS Number:
878144
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.07 (vs air)

vapor pressure

20 mmHg ( 30 °C)

assay

≥98.0% (GC)

autoignition temp.

518 °F

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.417 (lit.)

bp

117-118 °C/733 mmHg (lit.)

density

0.874 g/mL at 25 °C (lit.)

format

neat

SMILES string

COCCCN

InChI

1S/C4H11NO/c1-6-4-2-3-5/h2-5H2,1H3

InChI key

FAXDZWQIWUSWJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

80.6 °F - closed cup

flash_point_c

27 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service