Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10497

Sigma-Aldrich

Tungsten boride

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WB
CAS Number:
Molecular Weight:
194.65
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥97.0%

form

powder

reaction suitability

reagent type: catalyst
core: tungsten

SMILES string

B#[W]

InChI

1S/B.W

InChI key

JEEHQNXCPARQJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hasan Türkez et al.
Chemico-biological interactions, 273, 257-265 (2017-07-02)
During the recent years, microarray analysis of gene expression has become an inevitable tool for exploring toxicity of drugs and other chemicals on biological systems. Therefore, toxicogenomics is considered as a fruitful area for searching cellular pathways and mechanisms including

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service