Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08913

Sigma-Aldrich

(S)-4-Phenyloxazolidine-2-thione

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NOS
CAS Number:
Molecular Weight:
179.24
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.0%

optical activity

[α]/D +77±3°, c = 0.2 in chloroform

optical purity

enantiomeric ratio: ≥99.0:1.0 (LC)

SMILES string

S=C1N[C@H](CO1)c2ccccc2

InChI

1S/C9H9NOS/c12-9-10-8(6-11-9)7-4-2-1-3-5-7/h1-5,8H,6H2,(H,10,12)/t8-/m1/s1

InChI key

LVIJIGQKFDZTNC-MRVPVSSYSA-N

Application

A highly selective and efficient chiral auxiliary which can be directly reduced to its corresponding aldehyde and the chiral auxiliary by reductive cleavage with diisobutylaluminum hydride.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M T Crimmins et al.
Organic letters, 2(6), 775-777 (2001-02-07)
[formula: see text] Asymmetric aldol additions using chlorotitanium enolates of thiazolidinethione propionates proceed with high diastereoselectivity for the "Evans" or "non-Evans" syn product depending on the nature and amount of the base used. With (-)-sparteine as the base, selectivities of
Velazquez. F.; Olivo, H. F.
Current Organic Chemistry, 6, 303-303 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service