Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07691

9-cis,11-trans-Octadecadienoic acid solution

Synonym(s):

9Z,11E-CLA, Bovinic acid, Conjugated linoleic acid (9Z,11E)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O2
CAS Number:
Molecular Weight:
280.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

storage temp.

2-8°C

SMILES string

CCCCCC\C=C\C=C/CCCCCCCC(O)=O

InChI

1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h7-10H,2-6,11-17H2,1H3,(H,19,20)/b8-7+,10-9-

InChI key

JBYXPOFIGCOSSB-GOJKSUSPSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jason R Deguire et al.
Nutrition research (New York, N.Y.), 32(12), 911-920 (2012-12-19)
The relationships between conjugated linoleic acid (CLA) status, bone, body composition, and the effect of CLA on calciotropic hormones are unclear. A cross-sectional study was designed to examine the association between c9, t11 CLA status in erythrocyte membranes (RBC) and
M Joy et al.
Meat science, 90(3), 775-782 (2011-12-01)
The aim of this study was to evaluate the use of forage diets (grazing vs. hay) around the time of ewe parturition, on the fatty acids profile of suckling lamb meat (10-12kg body weight). Forty-eight multiparous single-bearing ewes were used.
Xiaohua Liu et al.
Journal of agricultural and food chemistry, 60(38), 9705-9710 (2012-09-06)
A simple and accurate method for screening of bacterial strains with the ability to convert free linoleic acid into specific conjugated linoleic acid (CLA) isomers has been developed by combining the ultraviolet spectral scan and capillary electrophoresis analysis. The ultraviolet
Clare M Reynolds et al.
The Journal of nutritional biochemistry, 24(2), 401-411 (2012-05-26)
Conjugated linoleic acid (CLA) is found naturally in meat and dairy products, and represents a potential therapeutic functional nutrient. However, given the discrepancies in isomer composition and concentration, controversy surrounds its proposed antidiabetic, antiobesity effects. This study focused on the
Eva Katharina Richter et al.
Lipids, 47(2), 161-169 (2011-08-13)
This study explores the potential use of stable carbon isotope ratios (δ(13)C) of single fatty acids (FA) as tracers for the transformation of FA from diet to milk, with focus on the metabolic origin of c9,t11-18:2. For this purpose, dairy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service