Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07319

Sigma-Aldrich

1-Butyl-3-methylimidazolium nitrate

≥95.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15N3O3
CAS Number:
Molecular Weight:
201.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥95.0% (HPLC)

form

crystals

impurities

≤1.0% water

functional group

nitrate

SMILES string

[O-][N+]([O-])=O.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.NO3/c1-3-4-5-10-7-6-9(2)8-10;2-1(3)4/h6-8H,3-5H2,1-2H3;/q+1;-1

InChI key

WPHIMOZSRUCGGU-UHFFFAOYSA-N

General description

1-Butyl-3-methylimidazolium nitrate is a hydrophilic room temperature ionic liquid[1] with low crystallization temperature.[2] It can be prepared from 1-butyl-3-methylimidazolium bromide by reacting with silver nitrate.[2]

Caution

may contain few white precipitates

pictograms

Flame over circleExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Ox. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water effect on physicochemical properties of 1-butyl-3-methylimidazolium based ionic liquids with inorganic anions.
Grishina EP, et al.
Journal of Molecular Liquids, 177, 267-272 (2013)
Bojana Katana et al.
Physical chemistry chemical physics : PCCP, 22(42), 24764-24770 (2020-10-28)
Ion specific effects of ionic liquid (IL) constituents on the surface charge and aggregation properties of two types of particles (positively charged amidine (AL) and polyimidazolium-functionalized sulfate (SL-IP-2) latexes) were investigated in IL solutions containing different anions and the 1-butyl-3-methylimidazolium
Preparation of 1-butyl-3-methylimidazolium salts and study of their phase behavior and intramolecular intractions.
Gruzdev MS, et al.
Russ. J. Gen. Chem., 79(8), 1720-1727 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service