Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05539

Supelco

Perchlorate ionophore I

Selectophore, function tested

Synonym(s):

Dichloro-tetraphenylporphyrinato-phosphorus(V)-chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H28Cl3N4P
CAS Number:
Molecular Weight:
750.05
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

quality

function tested

λmax

676 mm

SMILES string

[Cl-].Cl[P+]1(Cl)n2c3ccc2c(-c4ccccc4)c5ccc(n5)c(-c6ccccc6)c7ccc(c(-c8ccccc8)c9ccc(n9)c3-c%10ccccc%10)n17

InChI

1S/C44H28Cl2N4P.ClH/c45-51(46)49-37-25-26-38(49)43(31-17-9-3-10-18-31)35-23-24-36(48-35)44(32-19-11-4-12-20-32)40-28-27-39(50(40)51)42(30-15-7-2-8-16-30)34-22-21-33(47-34)41(37)29-13-5-1-6-14-29;/h1-28H;1H/q+1;/p-1/b41-33-,41-37-,42-34-,42-39-,43-35-,43-38-,44-36-,44-40-;

InChI key

XSEKVURIGRVPIT-WVGKTRQHSA-M

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

may contain ~1 mol dichloromethane

Other Notes

Determination of perchlorate in water and human urine[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perchlorate selective membrane electrodes based on a phosphorus(V)-tetraphenylporphyrin complex.
M. Shamsipur et al.
Sensors and Actuators B, Chemical, 89, 9-14 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service