Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04109

Sigma-Aldrich

N,N′-Dimethoxy-N,N′-dimethyloxamide

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH3)NCOCON(CH3)OCH3
CAS Number:
Molecular Weight:
176.17
Beilstein/REAXYS Number:
5330464
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

≥97.0%

mp

89-95 °C

SMILES string

CON(C)C(=O)C(=O)N(C)OC

InChI

1S/C6H12N2O4/c1-7(11-3)5(9)6(10)8(2)12-4/h1-4H3

InChI key

OXSWGKZLVLVHIS-UHFFFAOYSA-N

Other Notes

A useful α-keto amide and 1,2-diketone synthon. [1][2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.P. Sibi et al.
Tetrahedron Letters, 33, 1941-1941 (1992)
The Journal of Organic Chemistry, 60, 5016-5016 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service