Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03968

Sigma-Aldrich

Fuchsin solution

denaturated with 2-butanone, 2% in ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein/REAXYS Number:
4166684
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

quality

denaturated with 2-butanone

concentration

2% in ethanol

density

0.80 g/mL at 20 °C

suitability

suitable for microscopy

SMILES string

Br.CO[C@@H]1CCCN[C@H]1CC(C)=O

InChI

1S/C9H17NO2.BrH/c1-7(11)6-8-9(12-2)4-3-5-10-8;/h8-10H,3-6H2,1-2H3;1H/t8-,9+;/m0./s1

InChI key

DVSQBUUDEVNABC-OULXEKPRSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service