Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-0570

Sigma-Aldrich

Semicarbazide hydrochloride

SAJ special grade, ≥99.0%

Synonym(s):

N-Aminourea hydrochloride, Hydrazine carboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONHNH2 · HCl
CAS Number:
Molecular Weight:
111.53
Beilstein/REAXYS Number:
3593642
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
assay:
≥99.0%
grade:
SAJ special grade
form:
powder
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥99.0%

form

powder

availability

available only in Japan

mp

175-177 °C (dec.) (lit.)

SMILES string

Cl.NNC(N)=O

InChI

1S/CH5N3O.ClH/c2-1(5)4-3;/h3H2,(H3,2,4,5);1H

InChI key

XHQYBDSXTDXSHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Oral

target_organs

Bone

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janaina M Zanoveli et al.
Brain research, 1294, 106-115 (2009-08-04)
Serotonin (5-HT) plays a key role in the neural circuitry mediating unconditioned and conditioned fear responses related to panic and generalized anxiety disorders. The basolateral nucleus of the amygdala (BLA) and the dorsal periaqueductal gray (dPAG) appear to be mainly
Fumiko Marttila-Ichihara et al.
Journal of immunology (Baltimore, Md. : 1950), 184(6), 3164-3173 (2010-02-16)
Vascular adhesion protein-1 (VAP-1) is an endothelial, cell surface-expressed oxidase involved in leukocyte traffic. The adhesive function of VAP-1 can be blocked by anti-VAP-1 Abs and small-molecule inhibitors. However, the effects of VAP-1 blockade on antitumor immunity and tumor progression
Yuping Chen et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(9), 1668-1673 (2011-06-16)
The novel biotransformation of an aminopyrrolidine to an aminopiperidine during the metabolism of 5-(4-chlorophenyl)-3-methyl-2-((2R)-2-(((1-methylethyl)amino)methyl)-1-pyrrolidinyl)-6-(4-pyridinyl)-4(3H)-pyrimidinone (AMG657417) was investigated using the NADPH-fortified S9 fraction from human liver. The major metabolite (M18) had a protonated molecule (MH(+) m/z 438) identical to that of
Jing Ye et al.
Journal of agricultural and food chemistry, 59(17), 9313-9318 (2011-07-27)
Azodicarbonamide, as a bleaching agent and improving agent, is a permitted food additive in certain countries and can be determined by high-performance liquid chromatography. However, it partially degrades with the heat of processing to form trace amounts of semicarbazide, which
Zining Cui et al.
Molecules (Basel, Switzerland), 15(6), 4267-4282 (2010-07-27)
In order to find novel chitin synthesis inhibitors (CSIs) with good activity, benzoylphenylurea, a typical kind of CSIs, was chosen as the lead compound and 15 novel derivatives containing furan moieties were designed by converting the urea linkage of benzoylphenylureas

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service