Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FXA-RO

Roche

Restriction Protease Factor Xa

from bovine plasma

Synonym(s):

factor xa, restriction protease, protease factor xa

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

form

lyophilized

mol wt

(Mr = 43 kD)

packaging

pkg of 3 × 100 μg (11585924001)
pkg of 3 × 250 μg (11179896001)

manufacturer/tradename

Roche

optimum pH

8.3

shipped in

wet ice

storage temp.

2-8°C

General description

Special quality for the cleavage of fusion proteins.
Lyophilizate

Specificity

The coagulation factor Xa is a serine protease which recognizes the amino acid sequence Ile—Glu—Gly—Arg ↓ with a high degree of specificity. Cleavage of this sequence activates the natural substrate prothrombin to thrombin. This sequence can also be used as a restriction cleavage site for processing of recombinant fusion proteins. When a lack of specificity is due to steric hindrance, the introduction of an additional amino acid sequence can sometimes overcome the problem.

Application

Processing of fusion proteins at definite cleavage site. The restriction protease factor Xa is a serine protease which recognizes the amino acid sequence Ile — Glu — Gly — Arg ↓ with a high degree of specificity. This sequence may also be used as a restriction cleavage site for processing of recombinant fusion proteins.

Quality

Purity: highly purified (SDS-PAGE); no albumin added
Quality control: function tested
Inhibitors: APMSF, antithrombin III-heparin, DFP, PMSF, soybean trypsin inhibitor

Preparation Note

Working concentration: 1:10 to 1:200 (w/w)
A useable working concentration has a substrate ratio of 1:10 to 1:200 (w/w).

Note: The contents of a vial may be used for several simultaneous digests. When repeating the digest, a new vial should be used. Following this procedure ensures reproducibility and prevents possible contamination.
Storage conditions (working solution): A solution in redistilled water can be used for up to one week when stored at 4 °C.
An aqueous solution, stored in aliquots at -20 °C is stable for approx. one month.
Freezing aliquots increases enzyme stability in solution.

Other Notes

For life science research only. Not for use in diagnostic procedures.

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service