Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11464752001

Roche

Alkaline Phosphatase (AP), activated

from calf intestine

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204

biological source

bovine (calf) intestine

assay

>95% (HPLC)

form

lyophilized (clear, colorless solution after reconstitution)

usage

sufficient for ≤5 labeling reactions

specific activity

8 units/μg protein (At 25 °C with 4-nitro-phenylphosphate as substrate (pH 9.8)

)

packaging

pkg of 10 mg (approx. 40 mg lyophilizate)

manufacturer/tradename

Roche

shipped in

wet ice

storage temp.

2-8°C

General description

Orthophosphoric-monoester phosphohydrolase (alkaline optimum)
The quantity of enzyme is sufficient to conjugate approx. 2 mg immunoglobulin G (IgG). Roche recommends dividing the total quantity to be conjugated into 5 portions; each portion provides 0.3 to 0.5 ml of conjugate from approx. 0.35 mg IgG; this quantity can be diluted 1:4,000 to 1:10,000 for ELISA applications.

Contents
Alkaline phosphatase from calf intestine lyophilizate from 0.5 ml.

Application

The reagent can be used for labeling water-soluble substances with reactive and accessible primary amino groups (e.g., peptides or proteins) with alkaline phosphatase for use in analytical methods.
It is particularly suitable for the coupling of antibodies with alkaline phosphatase, as the resulting conjugate is optimal for use in immunochemical detection systems, for example,  ELISA, immunohistochemistry, and immunoblotting procedures.
The enzyme can be used without pre-activation for the labeling of Ig, Ig Fab, and Ig F(ab′)2 fragments from rabbit, mouse, sheep, and goat.

Quality

Purity: >95% (HPLC)

Preparation Note

Working solution: Dissolving the lyophilizate in 0.5 ml double dist. Water produces an alkaline phosphatase concentration of 20 mg/ml.
Storage conditions (working solution): The reconstituted solution is stable for 5 days at 2 to 8 °C. The solution can be aliquoted and shock frozen at -70 °C or below, and then stored at -15 to -25 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanne M M Hensen et al.
Methods in molecular biology (Clifton, N.J.), 1184, 295-308 (2014-07-23)
B cell epitope mapping is widely applied to determine antibody-binding sites. Several methods exist to map B cell epitopes and here we describe three methods that are characterized by the simultaneous analysis of multiple peptides. In the first approach a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service