Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE1124

Sigma-Aldrich

Anti-XPA Antibody, clone 1XPA Antibody-1E11

ascites fluid, clone 1XPA-1E11, from mouse

Synonym(s):

DNA repair protein complementing XP-A cells, XPA, Xeroderma pigmentosum group A-complementing protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1XPA-1E11, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... XPA(7507)

General description

DNA repair protein complementing XP-A cells (UniProt P23025; also known as Excision repair-controlling, Mutant xeroderma pigmentosum complementation group A, Xeroderma pigmentosum group A-complementing protein) is encoded by the XPA (also known as XP1, XPAC) gene (Gene ID 7507) in human. DNA lesions caused by UV irradiation, drugs, or other environmental factors are eliminated by two nucleotide excision repair (NER) pathways, Global genome repair (GGR) and transcription-coupled repair (TCR). In GGR, the removal of lesions requires their recognition by the repair factor XPC/HR23b and the subsequent opening of the DNA duplex by transcription factor II human (TFIIH). The resulting single-stranded structure is stabilized by XPA and replication protein A (RPA). XPG is recruited through its interaction with TFIIH on the 3′ side of the lesion and its positioning on the cut site requires RPA. The interaction between XPA and XPB (ERCC1) stimulates the recruitment of ERCC1-XPF on the 5′ side of the DNA lesion. The damaged oligonucleotide can then be removed through the double incision by XPG and ERCC1-XPF endonucleases. In TCR, these factors (except XPC/HR23B) are recruited by the stalled RNA pol II in front of the damage with the help of the CSB and CSA proteins.

Immunogen

Epitope: Near C-terminus
Linear peptide corresponding to human XPA sequence near C-terminus.

Application

Anti-XPA Antibody, clone 1XPA Antibody-1E11 is an antibody against XPA for use in Western Blotting, Immunocytochemistry.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
Western Blotting Analysis: A representative lot detected the ATP-dependent recruitment of XPA in HeLa nuclear extract onto the stalled RNA pol IIO on mono-cisplatin-damaged DNA (Riedl, T., et al. (2003). EMBO J. 22(19):5293-5303; Laine, J.P., and, Egly, J.M. (2006). EMBO J. 25(2):3873-97).
Immunocytochemistry Analysis: A representative lot detected XPA recruitment to the DNA damage sites in the nuclei of UV-irradated HeLa cells (Alekseev, S., et al. (2014). Chem Biol. 21(3):398-407).

Quality

Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected XPA in 10 µg of HeLa nuclear extract.

Target description

~37 kDa observed. Target band appears larger than the calculated molecular weight of 31.4 kDa, most likely due to posttranslational modifications. Uncharacterized band(s) may appear in some lysates.

Physical form

Mouse monoclonal IgG1κ ascites with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas H Sanderson et al.
PloS one, 8(11), e78627-e78627 (2013-11-14)
Recent advancements in isolation techniques for cytochrome c (Cytc) have allowed us to discover post-translational modifications of this protein. We previously identified two distinct tyrosine phosphorylated residues on Cytc in mammalian liver and heart that alter its electron transfer kinetics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service