Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HAEG047A0

Millipore

3mm Hydrophobic Edge Membrane 0.45µm 47mm white gridded autoclave packed

Filling machine operations, sterility testing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.11

material

gridded filter
mixed cellulose esters (MCE) membrane
white filter

sterility

non-sterile

feature

autoclavable
hydrophobic

packaging

box of 10 envelopes containing 10 pak

border W

3 mm , hydrophobic edge

filter diam.

47 mm

pore size

0.45 μm pore size

input

sample type wine
sample type cosmetics
sample type soft drink
sample type pharmaceutical(s)
sample type bottled water
sample type beverage(s)
sample type sport drink
sample type beer
sample type food(s)
sample type cider

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical
sterility testing

shipped in

ambient

Packaging

Autoclave pack (requires autoclaving) of 10 resealable envelopes containing each 10 filters and 10 pads

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service