Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX1697F

Supelco

Coulomat CK

for use with AQUASTAR®

Synonym(s):

Coulometric Cathode Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601

form

liquid

Quality Level

compatibility

for use with AQUASTAR®

shipped in

ambient

storage temp.

room temp

General description

AQUASTAR®

Application

AQUASTAR
AX1697F-1 is packaged in 20/400 glass bottle
AX1697F-31 is packaged in 38/430 glass bottle

Analysis Note

Color: Reddish to pale yellow
Form: Liquid
Performance: To pass test
Water (H2O): 1-50ppm

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service