Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AC111

Sigma-Aldrich

Sheep Anti-Mouse IgG Antibody, Species absorbed: Rat IgG

from sheep

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$213.00

$213.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$213.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$213.00


Check Cart for Availability

Request a Bulk Order

biological source

sheep

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

Based on ELISA analysis, this antibody reacts with whole molecule mouse IgG (IgG>>IgG2a>IgG>>>>IgG1) with minimal or no cross-reaction mouse IgA or IgM.
This antibody has been tested by ELISA and/or solid-phase absorbed to ensure minimal cross-reaction with Human IgG, Rabbit IgG, Rat IgG, Chicken IgY, Guinea Pig IgG, and Hamster IgG, but may cross-react with immunoglobulins from other species.

Immunogen

Purified antibody corresponding to mouse IgG.

Application

ELISA (Isotype) Analysis: A representative lot of this antibody detected whole molecule mouse IgG (IgG>>IgG2a>IgG>>>>IgG1) with minimal or no cross-reaction mouse IgA or IgM.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
Sheep Anti-Mouse IgG Antibody, Species absorbed: Rat IgG is an antibody against Sheep Mouse IgG for use in ELISA.

Quality

Evaluated by ELISA using IgG antibodies from various species.

ELISA Analysis: This antibody detected 200 ng of mouse IgG antibody among a panel of IgG antibodies from various species.

Polysorp® is a registered trademark of Thermo Fisher Scientific.

Target description

150 kDa calculated

Physical form

Format: Purified
Fractionated by Octanoic Acid and Ammonium Sulfate cut and then depleted with Rat IgG column and Mouse IgA column.
Purified sheep Polyclonal serum in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

PolySorp is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hayley Joseph et al.
Frontiers in immunology, 10, 1840-1840 (2019-08-27)
A long-standing challenge in malaria is the limited understanding of B cell immunity, previously hampered by lack of tools to phenotype rare antigen-specific cells. Our aim was to develop a method for identifying carbohydrate-specific B cells within lymphocyte populations and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service