Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABT251

Sigma-Aldrich

Anti-Arpin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Arp2/3 inhibition protein, Arpin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$429.00

$429.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$429.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$429.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ARPIN(348110)

General description

Arpin (UniProt Q7Z6K5; also known as Arp2/3 inhibition protein, UPF0552 protein C15orf38) is encoded by the ARPIN (also known as C15orf38) gene (Gene ID 348110) in human. Cell migration requires the generation of branched actin networks that power the protrusion of the plasma membrane in lamellipodia. The actin-related proteins 2 and 3 (Arp2/3) complex nucleates actin filaments by binding to the side of an existing filament and initiating branch formation. Arpin functions as a negative regulator of Arp2/3 activity, allowing cells to control directional persistence of migration by fine-tuning actin nucleation activity at the leading edge of the lamellipodium. Arpin depletion in both mammalian cells and Dictyostelium discoideum amoeba is reported to result in straighter trajectories. Arpin microinjection is demonstrated to induce fish keratocytes to turn.

Specificity

Reacts with both Arpin and the C15orf38-AP3S2 fusion (isoform 2) produced from a naturally occurring readthrough transcript.

Immunogen

Epitope: near Arpin C-terminus
KLH-conjugated linear peptide corresponding to human Arpin near the C-terminus.

Application

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected detected Arpin and C15orf38-AP3S2 in MEF-1 cells.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Arpin Antibody is validated for use in Western Blotting, Immunocytochemistry for the detection of Arpin.

Quality

Evaluated by Western Blotting in MEF-1 murine embryonic fibroblasts cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected detected Arpin and C15orf38-AP3S2 in 10 µg of human U2OS and murine MEF-1 embryonic fibroblasts cell lysate.

Target description

~28/42 kDa observed (24.9 kDa/Arpin and 43.9 kDa/C15orf38-AP3S2 calculated)

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gopinath M Sundaram et al.
The Journal of experimental medicine, 214(10), 2889-2900 (2017-08-23)
Epithelial carcinomas are well known to activate a prolonged wound-healing program that promotes malignant transformation. Wound closure requires the activation of keratinocyte migration via a dual-state molecular switch. This switch involves production of either the anti-migratory microRNA miR-198 or the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service