Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS1621

Sigma-Aldrich

Anti-Lcat Antibody

serum, from rabbit

Synonym(s):

Phosphatidylcholine-sterol acyltransferase, Lecithin-cholesterol acyltransferase, Phospholipid-cholesterol acyltransferase, Lcat

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$322.00

$322.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$322.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$322.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LCAT(3931)

General description

Phosphatidylcholine-sterol acyltransferase (EC 2.3.1.43; UniProt P16301; also known as Lecithin-cholesterol acyltransferase, Phospholipid-cholesterol acyltransferase, LCAT) is encoded by the Lcat (also known as AI046659; D8Wsu61e) gene (Gene ID 16816) in murine species. LCAT catalyzes the esterification of cholesterol and phosphatidylcholines (PCs) to cholesteryl esters and lysoPCs on the surface of high and low density lipoproteins (HDLs and LDLs). LCAT reaction is essential for the maturation of nascent HDL particles in plasma, and a genetic deficiency of LCAT results in the near absence of normal HDL particles and the presence of plasma very low density and low density lipoprotein particles of abnormal size and shape. LCAT is initially produced with a 24-aa N-terminal signal peptide, which is later cleaved to form the mature enzyme. LCAT is synthesized in the liver before it is secreted into the plasma.

Immunogen

Recombinant protein corresponding to mouse Lcat.

Application

This Anti-Lcat Antibody is validated for use in Western Blotting for the detection of Lcat .
Western Blotting Analysis: A representative lot detected Lcat in FPLC-fractionated, TCA-precipitated plasma HDL from wild-type mice as well as mice with human apoM and/or A-I transgene (Liu, M., et al. (2014). J Biol Chem. 289(5)2801-2814).
Western Blotting Analysis: A representative lot detected Lcat in plasma from WT and endothelial lipase (EL) knockout mice (Ma, K., et al. (2003). Proc Natl Acad Sci USA. 100(5)2748-2753).
Western Blotting Analysis: A representative lot detected purified murine Lcat and Lcat in plasma samples from human A-I transgenic mice with or without scavenger receptor class B type I (SR-BI) knockout, purified mouse LCAT (Lee, J.Y., et al. (2007). J Lipid Res. 48(5)1052-1061).

Quality

Evaluated by Western Blotting in mouse serum.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Lcat in ~2.3 µg of mouse serum.

Target description

~60 kDa observed. Target band size appears larger than the calculated molecular weight (49.7 kDa/preform & 49.0 kDa/mature) due to glycosylation. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service