ABE2888
Anti-phospho PPAR alpha (Ser73)
from rabbit
Synonym(s):
Peroxisome proliferator-activated receptor alpha, PPAR-alpha, Nuclear receptor subfamily 1 group C member 1
Select a Size
About This Item
Recommended Products
biological source
rabbit
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse
species reactivity (predicted by homology)
human (based on 100% sequence homology), bovine (based on 100% sequence homology), rat (based on 100% sequence homology)
technique(s)
dot blot: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
target post-translational modification
phosphorylation (pSer73)
Gene Information
human ... PPARA(5465)
General description
Specificity
Immunogen
Application
Epigenetics & Nuclear Function
Immunofluorescence Analysis: A representative lot detected phospho PPAR alpha (Ser73) in Immunofluorescence applications (Hinds, T.D., et. al. (2016). J Biol Chem. 291(48):25179-25191).
Immunocytochemistry Analysis: A representative lot detected phospho PPAR alpha (Ser73) in Immunocytochemistry applications (Hinds, T.D., et. al. (2016). J Biol Chem. 291(48):25179-25191).
Immunohistochemistry Analysis: A representative lot detected phospho PPAR alpha (Ser73) in Immunohistochemistry applications (Hinds, T.D., et. al. (2017). Am J Physiol Endocrinol Metab. 312(4):E244-E252).
Quality
Dot Blot Analysis: A 1:250 dilution from a representative lot detected phospho PPAR (Ser73) peptide.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service