Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1471

Sigma-Aldrich

Anti-phospho-DNA-dependent RNA Pol II, subunit B1 (Ser2/5) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

DNA-directed RNA polymerase II subunit RPB1, RNA polymerase II subunit B1, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase III largest subunit, RNA-directed RNA polymerase II subunit RPB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer2/pSer5)

Gene Information

human ... POLR2A(5430)

General description

DNA-directed RNA polymerase II subunit RPB1, also known as RNA polymerase II subunit 1, RNA polymerase II subunit B1, DNA-directed RNA polymerase III largest subunit, RNA polymerase II subunit B220, and encoded by the gene name RPO21, RPB1, RPB220, SUA8, is a member of the RNA polymerase beta chain family. DNA-dependendent RNA Pol II has been shown to catalyze the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. DNA-dependendent RNA Pol II is the largest and catalytic component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. It forms the polymerase active center together with the second largest subunit. Pol II is the central component of the basal RNA polymerase II transcription machinery. During a transcription cycle, Pol II, general transcription factors and the Mediator complex assemble as the preinitiation complex (PIC) at the promoter. Pol II is composed of mobile elements that move relative to each other. The core element with the central large cleft comprises RPB3, RBP10, RPB11, RPB12 and regions of RPB1 and RPB2 forming the active center. The clamp element (portions of RPB1, RPB2 and RPB3) is connected to the core through a set of flexible switches and moves to open and close the cleft. In elongating Pol II, the lid loop (RPB1) appears to act as a wedge to drive apart the DNA and RNA strands at the upstream end of the transcription bubble and guide the RNA strand toward the RNA exit groove located near the base of the largely unstructured CTD domain of RPB1. The rudder loop (RPB1) interacts with single-stranded DNA after separation from the RNA strand, likely preventing re-association with the exiting RNA. The cleft is surrounded by jaws: an upper jaw formed by portions of RBP1, RPB2 and RPB9, and a lower jaw, formed by RPB5 and portions of RBP1. The jaws are thought to grab the incoming DNA template, mainly by RPB5 direct contacts to DNA.

Immunogen

KLH-conjugated linear peptide corresponding to human phospho-DNA-dependent RNA Pol II, subunit B1 (Ser2/5) near the C-Terminus.

Application

Anti-phospho-DNA-dependent RNA Pol II, subunit B1 (Ser2/5) | ABE1471 is an antibody against phospho-DNA-dependent RNA Pol II for use in Western Blotting.

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho-DNA-dependent RNA Pol II, subunit B1 (Ser2/5) in 10 µg of NIH/3T3 cell lysate.

Target description

~220 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service