Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABE1384

Sigma-Aldrich

Anti-Histone H2B type 2-E (H2be) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H2B type 2-E, H2b 613, H2be, Histone 2 H2be

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$457.00

$457.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$457.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$457.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H2AC21(317772)
mouse ... H2Bc21(319190)

General description

Histone H2B type 2-E (UniProt Q64524; also known as H2b 613, Histone 2 H2be) is encoded by the Hist2h2be gene (Gene ID 319190) in murine species. H2be is a replication-independent histone H2b variant expressed exclusively in olfactory chemosensory neurons. Levels of H2be are heterogeneous among olfactory neurons, but stereotyped according to the identity of the co-expressed olfactory receptor (OR). H2be is nearly undetectable in highly active neurons, but almost completely replaces H2b in inactive neurons. Changes in H2be expression affect olfactory function and OR representation in the adult olfactory epithelium. Genetic studies show that the deletion of H2be significantly extends the lifespan of neurons, whereas elevated H2be shortens their lifespan.

Specificity

Broad species reactivity expected based on sequence similarity.
The identity of an immunoreactive band of ~16 kDa detected by this polyclonal antibody using HeLa cell lysate has not been determined, but most likely corresponds to human histone H2B type 1-L (Histone H2B.c; Uniprot Q99880) whose N-terminal sequence (up to a.a. 39) is identical to that of murine histone H2B type 2-E.

Immunogen

Epitope: N-terminal
KLH-conjugated linear peptide corresponding to the N-terminal sequence of mouse histone H2B type 2-E (H2be).

Application

Anti-Histone H2B type 2-E (H2be) Antibody is an antibody against Histone H2B type 2-E for use in Western Blotting, Dot Blot.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected H2B type 2-E (H2be) in brain tissue lysates from wild-type, but not from Hist2h2be-knock mice (Courtesy of Professor C. David Allis, The Rockefeller University, New York).
Dot Blot Analysis: Specificity of this polyclonal antibody was confirmed using the antiserum prior to purification. The antiserum detected the H2B type 2-E (H2be) immunogen peptide, but not peptides derived from human/murine/rat H2b and H3 N-terminal sequences (Courtesy of Professor C. David Allis, The Rockefeller University, New York).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected histone H2B type 2-E (H2be) in 10 µg of HeLa cell lysate.

Target description

~16 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service