Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB10527

Sigma-Aldrich

Anti-VDAC Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

voltage-dependent anion channel 1, Porin 31HL, Outer mitochondrial membrane protein porin 1, PORIN-31-HL, Porin 31HM, voltage-dependent anion-selective channel protein 1, Plasmalemmal porin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, pig, mouse, human, horse

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), bovine (based on 100% sequence homology), equine (based on 100% sequence homology), porcine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

Gene Information

human ... VDAC1(7416)

General description

Voltage-dependent anion-selective channel protein 1 (also known as VDAC, VDAC1 or outer mitochondrial membrane protein porin 1) is the the outer mitochondrial membrane receptor for hexokinase and BCL2L1. VDAC forms a channel through the mitochondrial membrane and is involved in small molecule diffusion, cell volume regulation and apoptosis. VDAC may participate in the formation of the permeability transition pore complex (PTPC), which is responsible for the release of mitochondrial products that triggers apoptosis.

Immunogen

KLH-conjugated linear peptide corresponding to human VDAC.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-VDAC Antibody is validated for use in WB for the detection of VDAC.

Quality

Evaluated by Western Blot in HepG2 cell lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected VDAC in 10 µg of HepG2 cell lysate.

Target description

~33 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HepG2 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanne M Hildebrand et al.
Nature communications, 11(1), 3150-3150 (2020-06-21)
MLKL is the essential effector of necroptosis, a form of programmed lytic cell death. We have isolated a mouse strain with a single missense mutation, MlklD139V, that alters the two-helix 'brace' that connects the killer four-helix bundle and regulatory pseudokinase
Khadija M Alawi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(10), 4285-4298 (2015-07-03)
Transient receptor potential vanilloid 1 (TRPV1) is involved in sensory nerve nociceptive signaling. Recently, it has been discovered that TRPV1 receptors also regulate basal body temperature in multiple species from mice to humans. In the present study, we investigated whether
Katerina Gkirtzimanaki et al.
Cell reports, 25(4), 921-933 (2018-10-26)
Interferon α (IFNα) is a prompt and efficient orchestrator of host defense against nucleic acids but upon chronicity becomes a potent mediator of autoimmunity. Sustained IFNα signaling is linked to pathogenesis of systemic lupus erythematosus (SLE), an incurable autoimmune disease
Kahli Zeitlow et al.
Biochimica et biophysica acta. Molecular basis of disease, 1863(11), 2973-2986 (2017-08-03)
A variable-length poly-T variant in intron 6 of the TOMM40 gene, rs10524523, is associated with risk and age-of-onset of sporadic (late-onset) Alzheimer's disease. In Caucasians, the three predominant alleles at this locus are Short (S), Long (L) or Very long
Girolamo Spagnoletti et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 40(4), 1010428318770957-1010428318770957 (2018-04-18)
Preoperative chemoradiation is currently the standard of care in locally advanced rectal carcinoma, even though a subset of rectal tumors does not achieve major clinically meaningful responses upon neoadjuvant chemoradiation. At present, no molecular biomarkers are available to predict response

Articles

Loading controls in western blotting application.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service