Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB10520

Sigma-Aldrich

Anti-DAP-5 Antibody

serum, from rabbit

Synonym(s):

Death-associated protein 5, aging-associated protein 1, eIF-4-gamma 2, eIF-4G 2, eIF4G 2, eukaryotic translation initiation factor 4 gamma, 2, eukaryotic translation initiation factor 4G-like 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$363.00

$363.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$363.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$363.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EIF4G2(1982)

General description

Death Associated Protein 5 (DAP-5) is a protein with high amino acid sequence homology to Eukaryotic Translation Initiation Factor 4G (eIF4G). Compared with eIF4G, DAP5 lacks the N-terminal region necessary for cap-dependent translation and has a unique C-terminal part functioning as a regulator for interferon-gamma induced cell death. DAP5 is cleaved at Asp790 during apoptosis . The C-terminal truncated form of DAP5 functions as a cap-independent translation initiation factor responsible for the mediation of its own translation during apoptosis.

Specificity

This antibody recognizes DAP-5.

Immunogen

Epitope: Unknown
KLH-conjugated linear peptide corresponding to human DAP-5.

application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-DAP-5 Antibody is validated for use in WB for the detection of DAP-5.

Quality

Evaluated by Western Blot on Jurkat cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected DAP-5 on 10 µg of jurkat cell lysate.

Target description

97 kDa

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Jurkat cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service