Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.51215

Sigma-Aldrich

Fmoc-Ser(tBu)-sulfamylbutyryl linker

Novabiochem®

Synonym(s):

Fmoc-Ser(tBu)-sulfamylbutyryl linker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32N2O8S
Molecular Weight:
532.61
UNSPSC Code:
12352209

form

powder

manufacturer/tradename

Novabiochem®

functional group

Fmoc

storage temp.

2-30°C

General description

The nucleophile-cleavable safety-catch sulfamylbutyryl linker pre-loaded with Fmoc-Ser(tBu). Used to prepare peptides containing C-terminal serine bearing amine and ester modifications.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service