Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65044C

Sigma-Aldrich

Hemacolor® Solution III

Harleco®, suitable for hematology

Synonym(s):

Hematology Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

form

liquid

Quality Level

manufacturer/tradename

Harleco®

mp

-0.1 °C

application(s)

diagnostic assay manufacturing
histology

hematology

shipped in

ambient

storage temp.

15-25°C

General description

HARLECO® Blue color reagent for fast staining in hematology and cytology

Application

HARLECO® Blue color reagent for fast staining in hematology and cytology
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany
HEMACOLOR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service