Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.05771

Sigma-Aldrich

4-DAMP

Synonym(s):

4-DAMP, 1,1-Dimethyl-4-diphenylacetoxypiper­idinium iodide, Muscarinic Receptor Antagonist, 4-DAMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26INO2
CAS Number:
Molecular Weight:
451.34
MDL number:
UNSPSC Code:
12352200

assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 100 mM
ethanol: 25 mM

storage temp.

2-8°C

InChI

1S/C21H26NO2.HI/c1-22(2)15-13-19(14-16-22)24-21(23)20(17-9-5-3-6-10-17)18-11-7-4-8-12-18;/h3-12,19-20H,13-16H2,1-2H3;1H/q+1;/p-1

InChI key

WWJHRSCUAQPFQO-UHFFFAOYSA-M

General description

An antagonist selective for mAChRs (Ki = 0.57, 7.3, 0.37, 0.72 and 0.55 nM at human M1, M2, M3, M4, and M5 receptors respectively). Used frequently in investigating the functional roles of M1 and M3 receptors in both central and peripheral nervous systems.

Biochem/physiol Actions

Primary Target
Muscarinic Receptors
Target Ki: 0.57, 7.3, 0.37, 0.72 and 0.55 nM at human M1, M2, M3, M4, and M5 receptors respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kruse, C., et al., 2012. Nature.482, 552.
Moriya, H., et al., 1999. Life Sci.64, 2351.
Ehlert, J., 1996. Life Sci.58, 1971.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service