Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

236012

Sigma-Aldrich

COX-2 Inhibitor II - CAS 181696-33-3 - Calbiochem

The COX-2 Inhibitor II, also referenced under CAS 181696-33-3, controls the biological activity of COX-2. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

COX-2 Inhibitor II - CAS 181696-33-3 - Calbiochem, SC-791, 4-[(5-Difluoromethyl-3-phenyl)-4-isoxazolyl]benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12F2N2O3S
CAS Number:
Molecular Weight:
350.34
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chloe D Rose et al.
Current biology : CB, 30(12), 2363-2373 (2020-05-11)
Adolescent idiopathic scoliosis (AIS) affects 3% to 4% of children between the ages of 11 and 18 [1, 2]. This disorder, characterized by abnormal three-dimensional spinal curvatures that typically develop during periods of rapid growth, occurs in the absence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service