Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01971

Sigma-Aldrich

Neo-Cytocolor Quick Staining Kit - for cytological samples

for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124

IVD

for in vitro diagnostic use

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

Storage temp.

15-25°C

General description

Neo-Cytocolor is a quick - 4 minute - staining kit based on the standard staining method acc. to Szczepanik for use in the analysis of cytological specimens. This kit dispenses with the use of xylene, containing the xylene substitute NEO-CLEAR instead. Standard fixatives can be used. Neo-Cytocolor is an IVD product. As it also is CE-certified, it can be applied in diagnostics in an accredited laboratory.The package is sufficient for approx. 1000 applications.
for microscopy

Application

For gynecological and clinical specimens. Contains Neo-Clear, xylene substitute

Analysis Note

Suitability for microscopy (Vaginal smear)passes testNucleiblue to dark violetCyanophilic cytoplasmblue-greenEosinophilic cytoplasmpinkkeratinized cytoplasmpink-orangeMicroorganismsblue-violetErythrocytesred

Legal Information

NEO-CYTOCOLOR is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service