Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S-020

Supelco

Surine Negative Urine Control

bottle of 50 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

packaging

bottle of 50 mL

manufacturer/tradename

Cerilliant®

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material is applicable for use in LC/MS or GC/MS applications for clinical chemistry, urine drug testing or forensic analysis. Special handling of this synthetic urine negative control is not required.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Surine is a trademark of Dyna-Tek Industries

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Optimized sample prep and chiral chromatography methods for the LC/MS analysis of these drug enantiomers in urine

In this study, optimized methods are presented for sample preparation and chiral chromatography for the LC/MS analysis of amphetamine and methamphetamine enantiomers in urine.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service