Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M-173

Supelco

Methylmalonic acid-13C4 solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4H6O4
CAS Number:
Molecular Weight:
122.06
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

O[13C]([13CH]([13C](O)=O)[13CH3])=O

InChI

1S/C4H6O4/c1-2(3(5)6)4(7)8/h2H,1H3,(H,5,6)(H,7,8)/i1+1,2+1,3+1,4+1

InChI key

ZIYVHBGGAOATLY-JCDJMFQYSA-N

General description

Methylmalonic acid is an indicator of vitamin B12 deficiency and methylmalonic acidemia. This internal standard is suitable for quantitation of methylmalonic acid levels by LC/MS or GC/MS in the diagnosis of vitamin B12 deficiency and methylmalonic acidemia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F

flash_point_c

2.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service