Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IMF-021

Supelco

iMethod Test Kit for NIDA 5 Drug Panel

kit of 16 × 1.0 mL ampules, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

packaging

kit of 16 × 1.0 mL ampules

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)
estupefaciente/psicótropo (Spain); Decreto Lei 15/93: Tabela IA/IIA/IB/IIB (Portugal)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

General description

A kit of certified reference solutions suitable for use with the NIDA 5 Drug Panel iMethod® application.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
iMethod is a registered trademark of CEM Corporation

Kit Components Also Available Separately

Product No.
Description
SDS

  • A-007(±)-Amphetamine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • A-0096-Acetylmorphine, 1.0 mg/mL solution, 1 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • B-004Benzoylecgonine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • C-006Codeine solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • M-005Morphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • M-009(±)-Methamphetamine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • P-007PCP (Phencyclidine) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • T-006(±)-11-nor-9-Carboxy-Δ9-THC solution, 100 μg/mL in methanol, ampule of 1 mL, Cerilliant®SDS

  • A-005(±)-Amphetamine-d5 (deuterium label on side chain) solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • A-0066-Acetylmorphine-d3 solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • B-001Benzoylecgonine-d3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • C-005Codeine-D3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • M-003Morphine-D3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • M-004(±)-Methamphetamine-D5 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • P-003PCP-D5 (Phencyclidine-D5) solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

  • T-004(±)-11-nor-9-Carboxy-Δ9-THC-D3 solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®SDS

See All (16)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service