Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G-025

Supelco

Gabapentin Impurity Mixture solution

0.84-1.4 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO
Molecular Weight:
153.22
UNSPSC Code:
41116107

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Gabapentin Impurity A
Diphenhydramine Impurity D, 1.4 mg/mL

Gabapentin Impurity B
Diphenhydramine Impurity D, 0.84 mg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

0.84-1.4 mg/mL in methanol

format

multi-component solution

storage temp.

−20°C

General description

Gabapentin is an antiepileptic drug, which is a structural analog of gamma-aminobutyric acid (GABA). It is used in the treatment of partial seizures in adults and children. This certified solution standard contains Gabapentin impurities including Impurity A, and B. This standard can be used for multiple HPLC applications including pharmaceutical testing, research, and Monograph testing methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

50.0 °F - closed cup

flash_point_c

10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service