ERD-033S
2,4-Dinitrotoluene solution
1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®
Synonym(s):
2,4-DNT
About This Item
Recommended Products
grade
certified reference material
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1.2 mL
manufacturer/tradename
Cerilliant®
concentration
1000 μg/mL in acetonitrile
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
format
single component solution
storage temp.
−20°C
InChI
1S/C7H6N2O4/c1-5-2-3-6(8(10)11)4-7(5)9(12)13/h2-4H,1H3
InChI key
RMBFBMJGBANMMK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
35.6 °F - closed cup
flash_point_c
2 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service