Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D-141

Supelco

(±)-cis-4,4′-Dimethylaminorex solution

1.0 mg/mL (1% 1M HCl in Methanol), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL (1% 1M HCl in Methanol)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

format

single component solution

storage temp.

−20°C

SMILES string

C[C@H]1[C@@H](C2=CC=C(C)C=C2)OC(N)=N1

InChI

1S/C11H14N2O/c1-7-3-5-9(6-4-7)10-8(2)13-11(12)14-10/h3-6,8,10H,1-2H3,(H2,12,13)/t8-,10-/m0/s1

InChI key

NPILLHMQNMXXTL-WPRPVWTQSA-N

General description

A Certified Spiking Solution® applicable for use in GC/MS or LC/MS testing applications in forensic analysis, clinical toxicology or urine drug testing. Also known by street names DMAR or Serotoni, (±)-cis-4,4′-Dimethylaminorex is a centrally acting sympathomimetic drug related to 4-methylaminorex and pemoline. The amphetamine analog has been sold as a novel psychoactive drug in illicit markets in both Europe and the US.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Met. Corr. 1 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anindita Dasgupta et al.
Nature communications, 12(1), 1180-1180 (2021-02-21)
3D single molecule localization microscopy (SMLM) is an emerging superresolution method for structural cell biology, as it allows probing precise positions of proteins in cellular structures. In supercritical angle localization microscopy (SALM), z-positions of single fluorophores are extracted from the
Anna M Van Keuren et al.
Cell reports, 33(10), 108486-108486 (2020-12-10)
The mitochondrial calcium uniporter is a multi-subunit Ca2+-activated Ca2+ channel, made up of the pore-forming MCU protein, a metazoan-specific EMRE subunit, and MICU1/MICU2, which mediate Ca2+ activation. It has been established that metazoan MCU requires EMRE binding to conduct Ca2+
Jennifer Martinez
Current protocols in cell biology, 87(1), e104-e104 (2020-05-22)
Phagocytes, notably macrophages, are critical sentinels of their environment, patrolling for and eradicating unwanted components. The ability of cells to process extracellular cargo in an appropriate manner is important for both clearance of the cargo and eventual return to homeostasis.
Travis B Kinder et al.
ACS chemical biology, 15(7), 1974-1986 (2020-05-28)
Immunosuppressants used to treat autoimmunity are often not curative and have many side effects. Our purpose was to identify therapeutics for autoimmunity of the skeletal muscle termed idiopathic inflammatory myopathies (myositis). Recent evidence shows that the pro-inflammatory type I interferons
Wenjun Wang et al.
Nature communications, 12(1), 476-476 (2021-01-22)
Endonuclease G (ENDOG), a mitochondrial nuclease, is known to participate in many cellular processes, including apoptosis and paternal mitochondrial elimination, while its role in autophagy remains unclear. Here, we report that ENDOG released from mitochondria promotes autophagy during starvation, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service