Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

840009P

Avanti

PAF (from Heart PC)

Avanti Research - A Croda Brand 840009P, powder

Synonym(s):

1-alkyl-2-acetoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (840009P-10mg)
pkg of 1 × 25 mg (840009P-25mg)
pkg of 1 × 5 mg (840009P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840009P

lipid type

phospholipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(C)=O)COCCCCCCCCCCCCCCCC)=O

InChI

1S/C12H26NO7P/c1-6-17-9-12(20-11(2)14)10-19-21(15,16)18-8-7-13(3,4)5/h12H,6-10H2,1-5H3/t12-/m1/s1

InChI key

OLGKYSVSAKLUHK-GFCCVEGCSA-N

General description

Platelet-activating factor (PAF; 1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine), an endogenous phospholipid is synthesized by several cells.[1] PAF is a phosphatidylcholine derivative.[2]

Biochem/physiol Actions

Platelet-activating factor (PAF; 1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine) is linked with arterial thrombosis, endotoxic shock, acute inflammation and acute allergic disease. It acts as a mediator of pathogenesis of ulcerative colitis and Crohn′s disease in human colon.[1] It also participates in platelet activation and inflammation.[2]

Packaging

5 mL Clear Glass Sealed Ampule (840009P-10mg)
5 mL Clear Glass Sealed Ampule (840009P-25mg)
5 mL Clear Glass Sealed Ampule (840009P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PAFAH Ib phospholipase A2 subunits have distinct roles in maintaining Golgi structure and function
Bechler ME and Brown WJ
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1831(3), 595-601 (2013)
Thromboxane A2-mediated Cl- secretion induced by platelet-activating factor in isolated rat colon
Suzuki T, et al.
European Journal of Pharmacology, 400(2-3), 297-303 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service