Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810738P

Avanti

16:0 TNP PE

Avanti Research - A Croda Brand 810738P, powder

Synonym(s):

1,2-Dipalmitoyl-sn-Glycero-3-Phosphoethanolamine-N-(2,4,6-trinitrophenyl) (Ammonium Salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C43H78N5O14P
Molecular Weight:
920.08
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810738P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810738P

lipid type

coenzymes
headgroup modified lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC1=C([N+]([O-])=O)C=C([N+]([O-])=O)C=C1[N+]([O-])=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]

General description

N-trinitrophenyl phosphatidylethanolamine (Tnp-PE) acts as a quencher of pyrene fluorescence.[1]

Packaging

5 mL Clear Glass Sealed Ampule (810738P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?-Cyclodextrins greatly enhance translocation of hydrophobic fluorescent phospholipids from vesicles to cells in culture IMPORTANCE OF MOLECULAR HYDROPHOBICITY IN PHOSPHOLIPID TRAFFICKING STUDIES
Tanhuanpa K & Somerharju P
The Journal of biological chemistry, 274(50), 35359-35366 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service