Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810246C

Avanti

16:0 TopFluor TMR Cardiolipin

Avanti Research - A Croda Brand, chloroform solution

Synonym(s):

1,1′,2,2′-Tetrapalmitoyl cardiolipin[3-(methoxyphenylpyrro-pyrrometheneboron difluoride)propanoyl], ammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C94H167BF2N4O19P2
CAS Number:
Molecular Weight:
1768.10
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mL (810246C-100UG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

100 μg/mL in chloroform (810246C-100UG)

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P([O-])(OC[C@]([H])(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)OCC(OC(CCC1=C(C)C2=CC3=CC=C(C4=CC=C(OC)C=C4)N3[B-](F)(F)[N+]2=C1C)=O)([H])COP([O-])(OC[C@]([H])(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O.[NH4+].[NH4+]

Packaging

5 mL Amber Glass Screw Cap Vial (810246C-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service