Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810230C

Avanti

C6-NBD Safingol

Avanti Research - A Croda Brand 810230C

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-L-threo-sphinganine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H51N5O6
CAS Number:
Molecular Weight:
577.76
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810230C-50ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810230C

concentration

0.05 mg/mL (810230C-50ug)

shipped in

dry ice

storage temp.

−20°C

General description

Safingol is a lysosphingolipid and a synthetic L-threo-stereoisomer of (D-erythro-) sphinganine.[1][2]

Biochem/physiol Actions

Safingol is a competitive inhibitor of sphingosine kinase 1 involved in cancer cell growth and proliferation. It is known to increase antitumor activity of cisplatin in xenograft experiments.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (810230C-50ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of threo-beta-aminoalcohols from aminoaldehydes via chelation-controlled additions. Total synthesis of l-threo sphingosine and safingol
Jung ME and Yi SW
Tetrahedron Letters, 53(32), 4216-4220 (2012)
Mark A Dickson et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 17(8), 2484-2492 (2011-01-25)
Sphingosine 1-phosphate (S1P) is an important mediator of cancer cell growth and proliferation. Production of S1P is catalyzed by sphingosine kinase 1 (SphK). Safingol, (l-threo-dihydrosphingosine) is a putative inhibitor of SphK. We conducted a phase I trial of safingol (S)
Jesse Coward et al.
Autophagy, 5(2), 184-193 (2008-12-23)
Safingol, the synthetic L-threo-stereoisomer of endogenous (D-erythro-) sphinganine, is an inhibitor of protein kinase C and sphingosine kinase in vitro, and in some cell types has been implicated in ceramide generation and induction of apoptosis. Utilizing electron microscopy, acridine orange

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service