MilliporeSigma
  • A phase I clinical trial of safingol in combination with cisplatin in advanced solid tumors.
A phase I clinical trial of safingol in combination with cisplatin in advanced solid tumors.
Clinical cancer research : an official journal of the American Association for Cancer Research (2011-01-25)
Mark A Dickson, Richard D Carvajal, Alfred H Merrill, Mithat Gonen, Lauren M Cane, Gary K Schwartz
ABSTRACT
Sphingosine 1-phosphate (S1P) is an important mediator of cancer cell growth and proliferation. Production of S1P is catalyzed by sphingosine kinase 1 (SphK). Safingol, (l-threo-dihydrosphingosine) is a putative inhibitor of SphK. We conducted a phase I trial of safingol (S) alone and in combination with cisplatin (C). A 3 + 3 dose escalation was used. For safety, S was given alone 1 week before the combination. S + C were then administered every 3 weeks. S was given over 60 to 120 minutes, depending on dose. Sixty minutes later, C was given over 60 minutes. The C dose of 75 mg/m(2) was reduced in cohort 4 to 60 mg/m(2) due to excessive fatigue. Forty-three patients were treated, 41 were evaluable for toxicity, and 37 for response. The maximum tolerated dose (MTD) was S 840 mg/m(2) over 120 minutes C 60 mg/m(2), every 3 weeks. Dose-limiting toxicity (DLT) attributed to cisplatin included fatigue and hyponatremia. DLT from S was hepatic enzyme elevation. S pharmacokinetic parameters were linear throughout the dose range with no significant interaction with C. Patients treated at or near the MTD achieved S levels of more than 20 μmol/L and maintained levels greater than and equal to 5 μmol/L for 4 hours. The best response was stable disease in 6 patients for on average 3.3 months (range 1.8-7.2 m). One patient with adrenal cortical cancer had significant regression of liver and lung metastases and another had prolonged stable disease. S was associated with a dose-dependent reduction in S1P in plasma. Safingol, the first putative SphK inhibitor to enter clinical trials, can be safely administered in combination with cisplatin. Reversible dose-dependent hepatic toxicity was seen, as expected from preclinical data. Target inhibition was achieved with downregulation of S1P. The recommended phase II dose is S 840 mg/m(2) and C 60 mg/m(2), every 3 weeks.