810225C
Avanti
18:1 Dansyl PS
Avanti Research™ - A Croda Brand 810225C
Synonym(s):
1,2-dioleoyl-sn-glycero-3-phospho-L-serine-N-(5-dimethylamino-1-naphthalenesulfonyl) (ammonium salt)
About This Item
Recommended Products
assay
>99% (TLC)
form
liquid
packaging
pkg of 1 × 1 mL (810225C-1mg)
pkg of 5 × 1 mL (810225C-5mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 810225C
concentration
1 mg/mL (810225C-1mg)
1 mg/mL (810225C-5mg)
shipped in
dry ice
storage temp.
−20°C
General description
Application
Biochem/physiol Actions
Packaging
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
target_organs
Liver,Kidney, Respiratory system
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
does not flash
flash_point_c
does not flash
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service