Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

810115P

Avanti

06:0-12:0 NBD PC

Avanti Research - A Croda Brand 810115P, powder

Synonym(s):

1-hexanoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H54N5O11P
CAS Number:
Molecular Weight:
715.77
UNSPSC Code:
12352211

form

powder

packaging

pkg of 1 × 1 mg (810115P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810115P

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Clear Glass Sealed Ampule (810115P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service