Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

550001P

Avanti

Aurora-DSG

Aurora-DSG Nanoparticles, powder

Synonym(s):

Gold-Lipid Conjugate; 1,2-distearoyl-sn-glycero-3-(Aurora Nanoparticle)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
Au55C262H265P12NO7
Molecular Weight:
14744.75
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>95% (TLC)

form

powder

packaging

pkg of 1 × 500 μg (with screw cap)

manufacturer/tradename

Avanti Research - A Croda Brand 550001P

shipped in

wet ice

storage temp.

2-8°C

General description

This new conjugate from Avanti consists of an Aurora gold cluster capped with triphenylphosphine ligands and a single diglyceride ligand. Fifty-five gold atom nanoparticles, first described by Schmid, are fascinating structures that are commonly used for contrast agents in various imaging applications. Ideally this particle, unlike larger colloidal style particles, is less intrusive to the target application and behaves like an organic entity instead of a foreign metallic probe. They also are a great generic platform for bottom up synthesis of novel, gold-centered nanostructures due to the ease and completeness of ligand exchange reactions. This complex is readily visible under TEM and can also be viewed in optical microscopes if further processed using a silver development procedure.

Biochem/physiol Actions

Aurora-DSG (Distearoylglycerol) nanoparticles aid in improving contrast in liposome and membrane preparations in cryo-transmission microscopy imaging. They are useful in lipid research as they provide good resolution image of the lipid bilayer on incorporation.[1] Their biophysical interaction with lipid bilayers is exploited in many membrane research studies. Confocal, atomic force and high-resolution microscopy utilize the incorporation of Aurora into the heterogeneous bilayers, as they do not hamper the lipid layer properties and composition.[2]

Packaging

5 mL PTFE Vial with Screw Cap (550001P-500ug)

Legal Information

Aurora is a trademark of Avanti Polar Lipids, LLC
Avanti Research is a trademark of Avanti Polar Lipids, LLC

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

target_organs

Central nervous system,Peripheral nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentina Marchesano et al.
ACS nano, 7(3), 2431-2442 (2013-03-02)
Gold nanoparticles have emerged as novel safe and biocompatible tools for manifold applications, including biological imaging, clinical diagnostics, and therapeutics. The understanding of the mechanisms governing their interaction with living systems may help the design and development of new platforms
Jesús Sot et al.
Chemistry and physics of lipids, 218, 40-46 (2018-12-07)
Lipid bilayers of different phospholipid compositions have been prepared, in the form of vesicles, or of supported lipid bilayers, and doped with Aurora™ at 0.1 mol%. Aurora™ consists of an Au55 gold nanoparticle (about 1.4 nm in diameter) capped with triphenylphosphine ligands
Wiktor Lewandowski et al.
Langmuir : the ACS journal of surfaces and colloids, 29(10), 3404-3410 (2013-02-21)
Hybrid structures made of metal nanoparticles with liquid crystalline coating attract considerable attention due to their conspicuous self-assembly and potential synergistic properties. Here we report on a new structural parameter that can be used to control the formation of hybrid
The interaction of lipid-liganded gold clusters (Aurora?) with lipid bilayers
Sot J, et al.
Chemistry and Physics of Lipids, 218(8), 40-46 (2019)
Ramjee Balasubramanian et al.
Journal of the American Chemical Society, 127(22), 8126-8132 (2005-06-02)
This paper describes the reaction of the phosphine-protected Au nanoparticle Au(55)(PPh(3))(12)Cl(6) (1, "Au55") with hexanethiol (2) and other thiols. The voltammetry of the reaction product 2 displays a well-defined pattern of peaks qualitatively reminiscent of Au(38) nanoparticles, but with quite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service