330021S
Avanti
WR304 mAb
Avanti Research™ - A Croda Brand 330021S
Synonym(s):
WR304 mAb, WR304 Mouse Monoclonal Antibody (IgM), Anti-(PIP/PIP2), 1X TBS (Tris Buffered Saline) with 1mM Na2 EDTA
Select a Size
Select a Size
About This Item
Recommended Products
antibody form
purified immunoglobulin
antibody product type
primary antibodies
assay
>95%
packaging
pkg of 1 × 0.1 mL (polypropylene tube with screw cap (330021S-100ug))
manufacturer/tradename
Avanti Research™ - A Croda Brand 330021S
concentration
1 mg/mL (330021S-100ug)
shipped in
dry ice
storage temp.
−70°C
General description
Formulation: WR304 is provided as a 0.2μm sterile-filtered solution in Tris buffered saline (TBS). WR304 binding to porcine brain PI(4)P is inhibited by high concentration of Ca2+ (10 mM)iv and addition of 1 mM EDTA stimulates this binding activity. PBS and other phosphate-containing buffers should not be used as they inhibit binding to the phospholipid head groupiii.
Specificity
Immunogen
Application
The WR304 antibody has been used for the quantitation of PIP/PIP2 in direct ELISA and Dot Blot analysis. The WR304 antibody specifically binds to PIP/PIP2 and inhibits the infectivity of HIV-1 to peripheral blood mononuclear cells (PBMC).
Packaging
Storage and Stability
Other Notes
Antibody Source: Monoclonal antibody from BALB/c-derived hybridoma WR304.
Production: In vitro cell culture. Purification: Ultra filtration through 100 KDa cut-off filters.
Purification: Ultra filtration through 100 KDa cut-off filters.
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service