Z655589
Hanna combination ISE electrodes
Potassium, Polymer membrane
All Photos(1)
About This Item
UNSPSC Code:
41115603
Recommended Products
material
polyetherimide body
manufacturer/tradename
Hanna HI 4114
ISE measuring range
39000-0.49 ppm
pH measuring range
2-12.0
parameter
0-40 °C temp. range
electrode immersion depth × O.D.
120 mm × 12 mm
General description
Determination of potassium ions in wine, waters, soils and biological samples.
These include a sensor and reference electrode in a single electrode body. Hanna combination ion selective electrodes provide selectivity and include superior double junction reference into an electrode body. Combination solid-state electrodes have a built-in solid-state sensor and quick refill reference electrode.
Hanna′s liquid membrane and fluoride combination electrodes have replaceable module construction and the Hanna double junction refence stability.
Hanna′s liquid membrane and fluoride combination electrodes have replaceable module construction and the Hanna double junction refence stability.
Choose from one of the most recent versions:
Certificates of Analysis (COA)