Key Documents
Z555541
Sigma-Aldrich
Food Chemicals Codex, 5th edition
About This Item
ISBN-10:
0-309-08866-6
ISBN-13:
978-0-309-08866-4
UNSPSC Code:
55101509
publication info
Committee on Food Chemicals Codex, National Academy Press, 2003, 1032 pp., hard cover
General description
The Food Chemicals Codex is the accepted standard for defining the quality and purity of food chemicals. It is frequently referenced by the U.S. Food and Drug Administration and many international food regulatory authorities. The Fifth Edition updates the field, incorporating the definitive monographs of previous editions with completely new material. Each monograph provides a physical description of the substance and its use in foods, lists its purity requirements, describes the appropriate tests to determine compliance, and provides packaging and storage guidelines.
Choose from one of the most recent versions:
Certificates of Analysis (COA)