Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W442000

Sigma-Aldrich

Ethyl isothiocyanate

97%

Synonym(s):

Ethyl mustard oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NCS
CAS Number:
Molecular Weight:
87.14
FEMA Number:
4420
Beilstein/REAXYS Number:
1737705
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

assay

97%

refractive index

n20/D 1.512 (lit.)

bp

130-132 °C (lit.)

mp

−6 °C (lit.)

density

0.995 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

garlic

SMILES string

CCN=C=S

InChI

1S/C3H5NS/c1-2-4-3-5/h2H2,1H3

InChI key

HBNYJWAFDZLWRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

75.2 °F - closed cup

flash_point_c

24 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James R Durig et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(3), 783-795 (2007-04-17)
Variable temperature (-105 to -150 degrees C) studies of the infrared spectra (3500-400 cm(-1)) of ethylisothiocyanate, CH(3)CH(2)NCS, dissolved in liquid krypton have been recorded. Additionally the infrared spectra of the gas and solid have been re-investigated. These spectroscopic data indicate
Hirokuni Tajima et al.
Bioscience, biotechnology, and biochemistry, 67(8), 1844-1846 (2003-09-03)
Hydroxy isothiocyanates, especially 2-(4-hydroxyphenyl)ethyl isothiocyanate (hITC), were examined for antimicrobial synergism with streptomycin (SM) against Escherichia coli. On the course of those experiments, a peculiar suppression of SM by a low concentration of hITC was observed, besides the antibacterial synergism
Antonietta Melchini et al.
Journal of medicinal chemistry, 55(22), 9682-9692 (2012-09-25)
Dietary isothiocyanates (ITC) derived from cruciferous vegetables have been shown to have numerous biological effects consistent with chemoprotective activity. In this study we synthesized a novel ITC, 2-(2-pyridyl) ethyl ITC (PY-ITC), and assessed its chemopreventive ability in comparison to sulforaphane
W H Mennicke et al.
Xenobiotica; the fate of foreign compounds in biological systems, 13(4), 203-207 (1983-04-01)
The metabolism of methyl, ethyl, butyl and allyl isothiocyanate, which occur as glucosinolates in a number of plants, was studied. Oral administration of the substances to the rat was followed by their renal excretion as mercapturic acids, which were isolated
Juyoung Lee et al.
Journal of the science of food and agriculture, 95(11), 2244-2251 (2014-10-02)
Glucosinolates are abundant in Brassicaceae vegetables, and they are degraded into various organic breakdown products (BPs) (R-CN, -NCS and -SCN) by myrosinase when plant tissues are damaged. This study was designed to investigate whether these BPs could be broken further

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service