Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

W314620

Sigma-Aldrich

2,2′-(Dithiodimethylene)difuran

≥96%, natural, FG

Synonym(s):

Furfuryl disulfide, Difurfuryl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2S2
CAS Number:
Molecular Weight:
226.32
FEMA Number:
3146
MDL number:
UNSPSC Code:
12352200
Flavis number:
13.050
organoleptic:
coffee; meaty; roasted; sulfurous
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
botanical
agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

botanical

grade

FG
Fragrance grade
Halal
Kosher
natural

agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.510

assay

≥96%

refractive index

n20/D 1.585 (lit.)

bp

112-115 °C/0.5 mmHg (lit.)

mp

10-11 °C (lit.)

density

1.233 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

organoleptic

coffee; meaty; roasted; sulfurous

SMILES string

C(SSCc1ccco1)c2ccco2

InChI

1S/C10H10O2S2/c1-3-9(11-5-1)7-13-14-8-10-4-2-6-12-10/h1-6H,7-8H2

InChI key

CBJPZHSWLMJQRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service