Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W283800

Sigma-Aldrich

Patchouli oil

Synonym(s):

Pogostemon cablin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2838
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Pogostemon cablin (Blanco) Benth

grade

Kosher

origin

Indonesia origin

refractive index

n20/D 1.509 (lit.)

bp

287 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

earthy; woody

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: steam distillation

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco Aguilar et al.
ACS omega, 5(50), 32436-32446 (2020-12-31)
Patchouli oil is a major ingredient in perfumery, granting a dark-woody scent due to its main constituent (-)-patchoulol. The growing demand for patchouli oil has raised interest in the development of a biotechnological process to assure a reliable supply. Herein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service