Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W231105

Sigma-Aldrich

Citronellyl acetate

mixture of isomers, natural, ≥95%, FG

Synonym(s):

3,7-Dimethyl-6-octen-1-yl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH2CH(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
198.30
FEMA Number:
2311
EC Number:
Council of Europe no.:
202
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.012
organoleptic:
lemon; rose
grade:
FG
Kosher
natural
food allergen:
no known allergens

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

assay

≥95%

refractive index

n20/D 1.445 (lit.)
n20/D 1.445

bp

240 °C (lit.)

density

0.888 g/mL at 25 °C
0.891 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

lemon; rose

SMILES string

CC(CCOC(C)=O)CC\C=C(\C)C

InChI

1S/C12H22O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,11H,5,7-9H2,1-4H3

InChI key

JOZKFWLRHCDGJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

217.4 °F - closed cup

flash_point_c

103 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyang-Sook Choi
Journal of agricultural and food chemistry, 53(5), 1642-1647 (2005-03-03)
This study was conducted to determine the composition of kumquat (Fortunella japonica Swingle) cold-pressed peel oil and to determine which volatile components are primarily responsible for the aroma of this oil. Eighty-two compounds were identified in the oil by GC
S Oda et al.
Bioscience, biotechnology, and biochemistry, 65(8), 1917-1919 (2001-10-02)
A double coupling system, which couples metabolism of glucose and transacetylation, is a unique procedure for the production of acetic esters. In the novel coupling system described in this article, acetyl coenzyme A (acetyl-CoA) was supplied via metabolism of both
Karin Förster-Fromme et al.
Microbiology (Reading, England), 154(Pt 3), 789-796 (2008-03-04)
The atuRABCDEFGH gene cluster is essential for acyclic terpene utilization (Atu) in Pseudomonas aeruginosa. The biochemical functions of most Atu proteins have not been experimentally verified; exceptions are AtuC/AtuF, which constitute the two subunits of geranyl-CoA carboxylase, the key enzyme
Jun Han et al.
Pest management science, 67(12), 1583-1588 (2011-06-16)
This study was aimed at assessing the fumigant toxicity of 14 essential oil constituents from lemon eucalyptus, Eucalyptus citriodora Hook, and another ten known compounds to females of acaricide-susceptible, chlorfenapyr-resistant, fenpropathrin-resistant, pyridaben-resistant and abamectin-resistant strains of Tetranychus urticae Koch. Menthol
H J Fang et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 24(5), 366-371 (1989-01-01)
Three geranium oils were separated, identified and compared by high resolution cross-linked fused silica capillary GC and GC-MSD. It was found that their chemical constituents are very similar, but the contents of some components (linalool, citronellol, cis-caryophyllene and guaiene) differ

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service