MilliporeSigma
  • [Studies on the chemical components and anti-tumour action of the volatile oils from Pelargonium graveoleus].
[Studies on the chemical components and anti-tumour action of the volatile oils from Pelargonium graveoleus].
Yao xue xue bao = Acta pharmaceutica Sinica (1989-01-01)
H J Fang, X L Su, H Y Liu, Y H Chen, J H Ni
ABSTRACT
Three geranium oils were separated, identified and compared by high resolution cross-linked fused silica capillary GC and GC-MSD. It was found that their chemical constituents are very similar, but the contents of some components (linalool, citronellol, cis-caryophyllene and guaiene) differ appreciably. Pharmacological experiments showed that citronellol, citronellyl formate, geraniol and citronellyl acetate exhibited marginal antitumour activities. The activity of the latter two components has not been reported in the literature.
Sigma-Aldrich
Citronellyl acetate, ≥95%, FCC, FG